missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GCC1 Protein
A DNA sequence encoding the Homo sapiens (Human) GCC1, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_078799.2 |
---|---|
Gene ID (Entrez) | 79571 |
Name | GCC1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16031701
|
enQuireBio™
QP7935-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16051701
|
enQuireBio™
QP7935-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16041701
|
enQuireBio™
QP7935-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16061701
|
enQuireBio™
QP7935-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16071701
|
enQuireBio™
QP7935-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16081701
|
enQuireBio™
QP7935-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_078799.2 | |
GCC1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
79571 | |
Research Use Only | |
GCC1 | |
Recombinant Protein | |
E. coli | |
FGMNFGGGPS KKDLLETIET QKKQLLQYQA RLKDVVRAYK SLLKEKEALE ASIKVLSVSH EADVGLAGVQ LPGLTFPDSV DDRCSTHSED STGTATSLDT AASLTSTKGE FGVEDDRPAR GPPPPKSEEA SWSESGVSSS SGDGPFAGGE VDKRLHQLKT QLATLTSSLA TVTQEKSRME ASYLADKKKM KQDLEDASNK AEEERARLEG ELKGLQEQIA ETKARLITQQ HDRAQEQSDH ALMLRELQKL LQEERTQRQD LELRLEETRE ALAGRAYAAE QMEGFELQTK QLTREVEELK SELQAIRDEK NQPDPRLQEL QEEAARLKSH FQAQLQQEMR KTALAEDQLR QQSQVEEQRV AALENQISEV SELLGTYEKA KQKDQLAIQK LKERILQLDL ENKTLALAAS SRSPLDSHGE ESSLDVNVLK DKMEKLKRLL QVAARKSQVT LDVEKLCDLE IMPSSEAADG EKATALYYQQ ELKQLKEEFE RYKMRAQVVL KSKNTKDGNL GKELEAAQEQ LAELKEKYIS LRLSCEELEH QHQQEADDWK QELARLQQLH RQELERCQLD FRDRTLKLEE ELHKQRDRAL AVLTEKDLEL EQLRSVALAS GLPGRRSPVG GGGPGDPADT SSSDSLTQAL QLAAANEPTF FLYAEQLARK EVEITSLRKQ KHRLEVEVHQ LQDRLLEEGE RHREEVAALQ SHIEKNIRDQ SREGANLEYL KNIIYRFLTL PDSLGRQQTL TAILTILHFS PEEKQVIMRL PTSASWWPSG KR | |
Greater than 90% as determined by SDS-PAGE. |