missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Fibulin 7 Protein

Product Code. 18725147
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18725147

Brand: R&D Systems™ 8530FB050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human Fibulin 7 Protein is derived from CHO. The Recombinant Human Fibulin 7 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q53RD9
For Use With (Application) Bioactivity
Gene ID (Entrez) 129804
Molecular Weight (g/mol) Observed M.W.: 56-65kDa, reducing conditions, Theoretical M.W.: 46kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Fibulin 7 protein Gln25-Phe439,with an N-terminal HA tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias FBLN7, FIBL-7, fibulin 7, TM14
Common Name TM14
Gene Symbol FBLN7
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of SVEC4-10 mouse vascular endothelial cells. The ED50 for this effect is typically 0.7-3.5μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived Gln25-Phe439, with an N-terminal HA tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.