missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human FGF-5 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with Trehalose and with BSA as a carrier protein.
Supplied as a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT.
Unit Size:
50µg
Description
The Recombinant Human FGF-5 Protein is derived from E. coli. The Recombinant Human FGF-5 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with Trehalose and with BSA as a carrier protein. |
| Gene ID (Entrez) | 2250 |
| Molecular Weight (g/mol) | 27 kDa |
| Name | FGF-5 |
| Quantity | 50 μg |
| Source | E. coli-derived human FGF-5 protein Glu23-Gly268 (Lys238Asn and Pro245Ser),with an N-terminal Met & Leu26-Gly268 (Lys238Asn and Pro245Ser) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction