missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human FABP9/T-FABP Protein

Product Code. 18739258
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity unitSize
18739258 100 μg 100µg
1 options
This item is not returnable. View return policy

Product Code. 18739258

Brand: R&D Systems™ 9488CL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Recombinant Human FABP9/T-FABP Protein CF is a member of a large superfamily of lipid binding proteins that are expressed in a tissue specific manner.

The Recombinant Human FABP9/T-FABP Protein is derived from E. coli.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q0Z7S8
Characterization Bioactivity not tested
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT.
Gene ID (Entrez) 646480
Molecular Weight (g/mol) 16kDa
Quantity 100 μg
Source E. coli-derived human FABP9/T-FABP protein Met1-Val132,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10EU per 1μg of the protein by the LAL method.
Gene Alias FABP-9, Fatty Acid Binding Protein 9, Testis, Fatty Acid-Binding Protein 9, Lipid-Binding Protein, PERF, PERF 15, PERF15, Testis Lipid Binding Protein, Testis Lipid-Binding Protein, Testis-Type Fatty Acid-Binding Protein, TFABP, T-FABP, TLBP
Product Type Recombinant Protein
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 1mg/mL in PBS.
Purity or Quality Grade >90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.