missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human DC-SIGN/CD209 Protein

Product Code. 18795057
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18795057 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18795057 Supplier R&D Systems™ Supplier No. 9136DC050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human DC-SIGN/CD209 Protein is derived from CHO. The Recombinant Human DC-SIGN/CD209 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NNX6
For Use With (Application) Bioactivity
Gene ID (Entrez) 30835
Molecular Weight (g/mol) Observed M.W.: 41-48kDa, reducing conditions, Theoretical M.W.: 41kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line,CHO-derived human DC-SIGN/CD209 protein, (N-terminus) YPYDVPDYA Human DC-SIGN/CD209 (Gln59-Ala404) Accession &Num; Q9NNX6 (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Common Name DC-SIGN/CD209
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of ICAM-3 expressing CHO Chinese hamster ovary cells. The ED50 for this effect is typically 0.4-2μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Chinese Hamster Ovary cell line, CHO-derived YPYDVPDYA, N-terminus; Human DC-SIGN/CD209 (Gln59-Ala404) Accession # Q9NNX6, C-terminus
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.