missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(XI) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XI) chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9074-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001177638.1 | |
Collagen alpha-1(XI) chain Protein | |
Research Use Only | |
XI | |
Recombinant Protein | |
Yeast | |
GPMGLTGRPG PVGGPGSSGA KGESGDPGPQ GPRGVQGPPG PTGKPGKRGR PGADGGRGMP GEPGAKGDRG FDGLPGLPGD KGHRGERGPQ GPPGPPGDDG MRGEDGEIGP RGLPGEAGPR GLLGPRGTPG APGQPGMAGV DGPPGPKGNM GPQGEPGPPG QQGNPGPQ | |
Greater than 90% as determined by SDS-PAGE. |
1301 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |