missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(XI) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XI) chain, was expressed in the hosts and tags indicated.
287.00€ - 3175.00€
Specifications
Accession Number | NP_001177638.1 |
---|---|
Gene ID (Entrez) | 1301 |
Name | Collagen alpha-1(XI) chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16057571
|
enQuireBio™
QP9074-YE-100UG |
100 μg |
920.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16097571
|
enQuireBio™
QP9074-YE-500UG |
500 μg |
2650.00€
500µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16067571
|
enQuireBio™
QP9074-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16087571
|
enQuireBio™
QP9074-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16007581
|
enQuireBio™
QP9074-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16077571
|
enQuireBio™
QP9074-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001177638.1 | |
Collagen alpha-1(XI) chain Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
1301 | |
Research Use Only | |
XI | |
Recombinant Protein | |
Yeast | |
GPMGLTGRPG PVGGPGSSGA KGESGDPGPQ GPRGVQGPPG PTGKPGKRGR PGADGGRGMP GEPGAKGDRG FDGLPGLPGD KGHRGERGPQ GPPGPPGDDG MRGEDGEIGP RGLPGEAGPR GLLGPRGTPG APGQPGMAGV DGPPGPKGNM GPQGEPGPPG QQGNPGPQ | |
Greater than 90% as determined by SDS-PAGE. |