missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CNTFR/CNTF Complex Protein
Click to view available options
Quantity:
20 μg
50 μg
Unit Size:
20µg
50µg
Description
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells.
Specifications
Specifications
| Accession Number | P26992 & P26441 |
| For Use With (Application) | Bioactivity |
| Molecular Weight (g/mol) | 59 kDa |
| Quantity | 20 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CNTFR/CNTF Complex protein Human CNTFR (Gln23-Ser342) Accession # P26992GGGSGGGSGGGSHuman CNTF (Met1-Met200) Accession # P26441N-terminus C-terminus |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Species | Human |
| Reconstitution | Reconstitute at 200 μg/mL in PBS. |
| Buffer | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction