missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human CLPP Protein
Shop All Bio Techne Products
Click to view available options
Unit Size:
250µg
Description
An un-tagged recombinant protein corresponding to the amino acids 57-277 of Human CLPP The Recombinant Human CLPP Protein is derived from E. coli. The Recombinant Human CLPP Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_006003 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 7.5) containing 2mM DTT, 20% glycerol, 100mM NaCl |
| Gene ID (Entrez) | 8192 |
| Molecular Weight (g/mol) | 24.2kDa |
| Name | CLPP Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | -80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >90% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction