missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CD30/TNFRSF8 Fc Chimera Biotin Protein, CF
Click to view available options
:
50 μg
Description
Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human CD30/TNFRSF8 Fc Chimera (Catalog # BT11155) binds to Recombinant Human CD30 Ligand/TNFSF8 (Catalog # 1028-CL ) with an ED50 of 0.800-16.0 ng/mL.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 943 |
| Molecular Weight (g/mol) | MolecularWeight-observed: 80-115 kDa, under reducing conditions., MolecularWeight-theroretical: 65 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human CD30/TNFRSF8 protein Human CD30/TNFRSF8 (Phe19-Lys379) (N-terminus) Accession # P28908.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | CD30, CD30 antigen, CD30KI-1, CD30L receptor, cytokine receptor CD30, D1S166EKi-1, Ki-1 antigen, Lymphocyte activation antigen CD30, TNFRSF8, tumor necrosis factor receptor superfamily member 8, tumor necrosis factor receptor superfamily, member 8 |
| Conjugate | Biotin |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction