missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL5 / RANTES Protein
A DNA sequence encoding the Homo sapiens (Human) CCL5 / RANTES, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8524-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002976.2 | |
CCL5 / RANTES Protein | |
Research Use Only | |
CCL5 | |
Recombinant Protein | |
E. coli | |
SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS | |
Greater than 90% as determined by SDS-PAGE. |
6352 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |