missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL5 / RANTES Protein
A DNA sequence encoding the Homo sapiens (Human) CCL5 / RANTES, was expressed in the hosts and tags indicated.
160.00€ - 1855.00€
Specifications
Accession Number | NP_002976.2 |
---|---|
Gene ID (Entrez) | 6352 |
Name | CCL5 / RANTES Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16025101
|
enQuireBio™
QP8524-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
16075101
|
enQuireBio™
QP8524-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16035101
|
enQuireBio™
QP8524-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16055101
|
enQuireBio™
QP8524-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16065101
|
enQuireBio™
QP8524-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16045101
|
enQuireBio™
QP8524-EC-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_002976.2 | |
CCL5 / RANTES Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6352 | |
Research Use Only | |
CCL5 | |
Recombinant Protein | |
E. coli | |
SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS | |
Greater than 90% as determined by SDS-PAGE. |