missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CCL5 / RANTES Protein

A DNA sequence encoding the Homo sapiens (Human) CCL5 / RANTES, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8524-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055101

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_002976.2
CCL5 / RANTES Protein
Research Use Only
CCL5
Recombinant Protein
E. coli
SPYSSDTTPC CFAYIARPLP RAHIKEYFYT SGKCSNPAVV FVTRKNRQVC ANPEKKWVRE YINSLEMS
Greater than 90% as determined by SDS-PAGE.
6352
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers