Learn More
R&D Systems™ Recombinant Human Caspase-10 (Mch 4) Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
The Recombinant Human Caspase-10 (Mch 4) Protein is derived from E. coli. The Recombinant Human Caspase-10 (Mch 4) Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in HEPES, NaCl and Sucrose. |
| Gene ID (Entrez) | 843 |
| Molecular Weight (g/mol) | 30 kDa |
| Name | Caspase-10 (Mch4) |
| Quantity | 10 μg |
| Source | E. coli-derived human Caspase-10 protein Val220-Ile479,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |