missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Casein kinase II subunit beta Protein

A DNA sequence encoding the Homo sapiens (Human) Casein kinase II subunit beta, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8381-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16074321

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

Casein kinase II subunit beta Protein
Research Use Only
CSNK2B
Recombinant Protein
E. coli
SSSEEVSWIS WFCGLRGNEF FCEVDEDYIQ DKFNLTGLNE QVPHYRQALD MILDLEPDEE LEDNPNQSDL IEQAAEMLYG LIHARYILTN RGIAQMLEKY QQGDFGYCPR VYCENQPMLP IGLSDIPGEA MVKLYCPKCM DVYTPKSSRH HHTDGAYFGT GFPHMLFMVH PEYRPKRPAN QFVPRLYGFK IHPMAYQLQL QAASNFKSPV KTIR
Greater than 90% as determined by SDS-PAGE.
50 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers