missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Casein kinase II subunit beta Protein
A DNA sequence encoding the Homo sapiens (Human) Casein kinase II subunit beta, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Name | Casein kinase II subunit beta Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | CSNK2B |
Protein Length | Partial (see sequence information for more details). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024321
|
enQuireBio™
QP8381-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16044321
|
enQuireBio™
QP8381-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16034321
|
enQuireBio™
QP8381-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16054321
|
enQuireBio™
QP8381-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16064321
|
enQuireBio™
QP8381-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16074321
|
enQuireBio™
QP8381-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
Casein kinase II subunit beta Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
CSNK2B | |
Recombinant Protein | |
E. coli | |
SSSEEVSWIS WFCGLRGNEF FCEVDEDYIQ DKFNLTGLNE QVPHYRQALD MILDLEPDEE LEDNPNQSDL IEQAAEMLYG LIHARYILTN RGIAQMLEKY QQGDFGYCPR VYCENQPMLP IGLSDIPGEA MVKLYCPKCM DVYTPKSSRH HHTDGAYFGT GFPHMLFMVH PEYRPKRPAN QFVPRLYGFK IHPMAYQLQL QAASNFKSPV KTIR | |
Greater than 90% as determined by SDS-PAGE. |