missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Calnexin Protein

Product Code. 18097044 Shop All Bio Techne Products
Click to view available options
:
0.1mg
This item is not returnable. View return policy

Product Code. 18097044

Brand: Novus Biologicals™ NBP130235

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results.

An un-tagged recombinant protein corresponding to the amino acids 21-481 of Human Calnexin The Recombinant Human Calnexin Protein is derived from E. coli. The Recombinant Human Calnexin Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P27824
For Use With (Application) ELISA, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH8.0), 1mM DTT, and 20% glycerol
Gene ID (Entrez) 821
Molecular Weight (g/mol) 52.5kDa
Name Calnexin Protein
Purification Method Protein
Quantity 0.1mg
Source E. Coli
Immunogen Calnexin, 21-481 aa, Human, Recombinant, E.coli (NP_001019820).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.