missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CALCA / CGRP Protein

A DNA sequence encoding the Homo sapiens (Human) CALCA / CGRP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8831-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16096711

  • 2095.00€ / 1mg

Estimated Shipment: 22-05-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001029125.1
CALCA / CGRP Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
796
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CALCA
Recombinant Protein
E. coli
ACDTATCVTH RLAGLLSRSG GVVKNNFVPT NVGSKA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers