missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human B7-H4 Fc Chimera Biotinylated Protein, CF

Product Code. 30485266
Click to view available options
:
50 μg
This item is not returnable. View return policy

Product Code. 30485266

Brand: R&D Systems™ BT11674050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human B7-H4 Fc Chimera binds to Human B7-H4 Antibody (Catalog # MAB6576 ) with an ED50 of 1.50-15.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q7Z7D3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 79679
Molecular Weight (g/mol) Observed: 75-95 kDa, under reducing conditions,Theoretical: 52 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human B7-H4 protein Human B7-H4 (Phe29-Ala258) (N-terminus) Accession # Q7Z7D3 Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias B7h.5, B7H4, B7-H4, B7H4T-cell costimulatory molecule B7x, B7S1, B7S1VCTN1, B7x, B7XPRO1291, FLJ22418, Immune costimulatory protein B7-H4, Protein B7S1, T cell costimulatory molecule B7x, V-set domain containing T cell activation inhibitor 1, V-set domain-containing T-cell activation inhibitor 1, Vtcn1
Conjugate Biotin
Reconstitution Reconstitute at 200 μg/mL in water.
Protein B7-H4
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.