Learn More
Novus Biologicals™ Recombinant Human Asprosin His Protein
Product Code. 18358366 Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. 18358366
Brand: Novus Biologicals™ NBP318164
Recombinant Protein
TRUSTED_SUSTAINABILITY
Specifications
Accession Number P35555
Concentration LYOPH
For Use With (Application) SDS-PAGE
Formulation Lyophilized from 0.2um-filtered solution in 50mM Tris-HCl (pH 8.2), 200mM NaCl
Gene ID (Entrez) 2200
Molecular Weight (g/mol) M.W. Theoretical: 18 kDa
Quantity 10 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <0.1 EU/μg purified protein (LAL test; Lonza).
Gene Alias FBN1, fibrillin 1
Gene Symbol FBN1
Reconstitution Reconstitute with 100 ul sterile water (After reconstitution: for 10 ug size: 0.1mg/ml)
Research Category Lipid and Metabolism
Purity or Quality Grade >95% pure by SDS-PAGE
Protein Asprosin