missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARNT Protein
A DNA sequence encoding the Homo sapiens (Human) ARNT, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8996-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001184254.1 | |
ARNT Protein | |
Research Use Only | |
ARNT | |
Recombinant Protein | |
Yeast | |
MAATTANPEM TSDVPSLGPA IASGNSGPGI QGGGAIVQRA IKRRPGLDFD DDGEGNSKFL RCDDDQMSND KERFARSDDE QSSADKERLA RENHSEIERR RRNKMTAYIT ELSDMVPTCS ALARKPDKLT ILRMAVSHMK SLRGTGNTST DGSYKPSFLT DQELKHLILE AADGFLFIVS CETGRVVYVS DSVTPVLNQP QSEWFGSTLY DQVHPDDVDK LREQLSTSEN ALTGRILDLK TGTVKKEGQQ SSMRMCMGSR RSFICRMRCG SSSVDPVSVN RLSFVRNRCR NGLGSVKDGE PHFVVVHCTG YIKAWPPAGV SLPDDDPEAG QGSKFCLVAI GRLQVTSSPN CTDMSNVCQP TEFISRHNIE GIFTFVDHRC VATVGYQPQE LLGKNIVEFC HPEDQQLLRD SFQQVVKLKG QVLSVMFRFR SKNQEWLWMR TSSFTFQNPY SDEIEYIICT NTNVKNSSQE PRPT | |
Greater than 90% as determined by SDS-PAGE. |
405 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |