missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ARNT Protein
A DNA sequence encoding the Homo sapiens (Human) ARNT, was expressed in the hosts and tags indicated.
202.00€ - 2155.00€
Specifications
Accession Number | NP_001184254.1 |
---|---|
Gene ID (Entrez) | 405 |
Name | ARNT Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16017431
|
enQuireBio™
QP8996-YE-100UG |
100 μg |
645.00€
100µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16057431
|
enQuireBio™
QP8996-YE-500UG |
500 μg |
1740.00€
500µg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16027431
|
enQuireBio™
QP8996-YE-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16047431
|
enQuireBio™
QP8996-YE-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16067431
|
enQuireBio™
QP8996-YE-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16037431
|
enQuireBio™
QP8996-YE-1MG |
1 mg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001184254.1 | |
ARNT Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
405 | |
Research Use Only | |
ARNT | |
Recombinant Protein | |
Yeast | |
MAATTANPEM TSDVPSLGPA IASGNSGPGI QGGGAIVQRA IKRRPGLDFD DDGEGNSKFL RCDDDQMSND KERFARSDDE QSSADKERLA RENHSEIERR RRNKMTAYIT ELSDMVPTCS ALARKPDKLT ILRMAVSHMK SLRGTGNTST DGSYKPSFLT DQELKHLILE AADGFLFIVS CETGRVVYVS DSVTPVLNQP QSEWFGSTLY DQVHPDDVDK LREQLSTSEN ALTGRILDLK TGTVKKEGQQ SSMRMCMGSR RSFICRMRCG SSSVDPVSVN RLSFVRNRCR NGLGSVKDGE PHFVVVHCTG YIKAWPPAGV SLPDDDPEAG QGSKFCLVAI GRLQVTSSPN CTDMSNVCQP TEFISRHNIE GIFTFVDHRC VATVGYQPQE LLGKNIVEFC HPEDQQLLRD SFQQVVKLKG QVLSVMFRFR SKNQEWLWMR TSSFTFQNPY SDEIEYIICT NTNVKNSSQE PRPT | |
Greater than 90% as determined by SDS-PAGE. |