Learn More
R&D Systems™ Recombinant Human ARC Western Blot Standard Protein
Product Code. 18156360
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 18156360
missing translation for 'mfr': R&D Systems™ WBC009
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
The Recombinant Human ARC Western Blot Standard Protein has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Gene ID (Entrez) 8996
Name ARC Western Blot Standard
Quantity 100 μL
Storage Requirements Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Conjugate Unconjugated
Recombinant Recombinant