missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Apoptosis regulatory protein Siva Protein

A DNA sequence encoding the Homo sapiens (Human) Apoptosis regulatory protein Siva, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6688-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095120

  • 560.00€ / 100µg

Estimated Shipment: 31-05-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_006418.2
Apoptosis regulatory protein Siva Protein
100 μg
Not determined.
Full Length of Isoform 2
Human
His
Tris-based buffer, 50% glycerol
10572
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SIVA1
Recombinant Protein
Yeast
MPKRSCPFAD VAPLQLKVRV SQRELSRGVC AERYSQEVFD PSGVASIACS SCVRAVDGKA VCGQCERALC GQCVRTCWGC GSVACTLCGL VDCSDMYEKV LCTSCAMFET
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers