Learn More
R&D Systems™ Recombinant Human Annexin A3 Protein
Product Code. 18794929
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18794929 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18794929 Supplier R&D Systems™ Supplier No. 9410AN050
 Restricted Item
Estimated Shipment: 04-12-2026
Calcium-dependent phospholipid-binding proteins.
The Recombinant Human Annexin A3 Protein is derived from E. coli.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Accession Number P12429
For Use With (Application) Flow Cytometry
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 306
Molecular Weight (g/mol) 37 kDa
Quantity 50 μg
Source E. coli-derived human Annexin A3 protein Ala2-Asp323,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method
Gene Alias 35-alpha calcimedin, annexin A3,1,2-cyclic-inositol-phosphate phosphodiesterase, Annexin III, annexin-3, ANXA3, Calcimedin 35-alpha Lipocortin III, PAP-III, Placental anticoagulant protein III, placental anticoagulant protein III, calcimedin 35-alpha
Product Type Recombinant protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain
Show More Show Less
For Research Use Only.