Learn More
R&D Systems™ Recombinant Human Annexin A1 Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Human Annexin A1 Protein is derived from E. coli. The Recombinant Human Annexin A1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 301 |
| Molecular Weight (g/mol) | 39.4 kDa |
| Name | Annexin A1 |
| Quantity | 50 μg |
| Source | E. coli-derived human Annexin A1 protein Met1-Asn346,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Monomer |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |