missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Angiopoietin-like 5 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS and NaCl.
Unit Size:
25µg
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human Angiopoietin-like Protein 5/ANGPTL5 (Catalog # 6675-AN/CF) is immobilized at 2 μg/mL (100 μL/well), Recombinant Human LILRB2/CD85d/ILT4 Fc Chimera (Catalog # 2078-T4 ) binds with an ED50 of 80.0-800 ng/mL.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS and NaCl. |
| Gene ID (Entrez) | 253935 |
| Molecular Weight (g/mol) | 42.3 kDa |
| Name | Angiopoietin-like Protein 5/ANGPTL5 |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human Angiopoietin-like Protein 5/ANGPTL5 protein Met1-Lys388 with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction