missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Aldo-keto Reductase 1B10/AKR1B10
Click to view available options
:
20ug; Carrier Free
Description
The Recombinant Human Aldo-keto Reductase 1B10/AKR1B10 is derived from Sf 21 (baculovirus). The Recombinant Human Aldo-keto Reductase 1B10/AKR1B10 has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, DTT and Glycerol. |
| Gene ID (Entrez) | 57016 |
| Molecular Weight (g/mol) | 37 kDa |
| Name | Aldo-keto Reductase 1B10/AKR1B10 |
| Quantity | 20μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Aldo-keto Reductase 1B10/AKR1B10 protein Ala2-Tyr316,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain at 5 μg per lane. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction