missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human aFGF / FGF1 Protein

Product Code. 16096701
Click to view available options
Quantity:
10 μg
100 μg
250 μg
50 μg
Unit Size:
100µg
10µg
250µg
50µg
This item is not returnable. View return policy

Product Code. 16096701

Brand: enQuireBio™ QP883250ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Human aFGF / FGF1 Protein, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_000791.1
Gene ID (Entrez) 2246
Name aFGF / FGF1 Protein
Quantity 250 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol FGF1
Protein Length Partial (see sequence information for more details).
Biological Activity The ED50 as determined by a cell proliferation assay using murine balb/c 3T3 cells is less than 0.5 ng/ml, corresponding to a specific activity of > 2.0 M 106 IU/mg in the presence of 10 ug/ml of heparin.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D
Buffer 0.2 um filtered PBS, pH 7.4, lyophilized
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.