Learn More
R&D Systems™ Recombinant Human Activin RIIA Fc Chimera Protein
Product Code. p-4719005
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
100µg
Product Code. 18350588
Brand: R&D Systems™ 340R2100/CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
The Recombinant Human Activin RIIA Fc Chimera Protein is derived from Sf 21 (baculovirus). The Recombinant Human Activin RIIA Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 92
Molecular Weight (g/mol) 40 kDa (monomer)
Name Activin RIIA Fc Chimera
Quantity 100 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Activin RIIA protein, (N-terminus) Human Activin RIIA (Ser25-Pro134) Accession &Num; P27037 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.