missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 28S ribosomal protein S6, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 28S ribosomal protein S6, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6386-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_115865.1 | |
28S ribosomal protein S6, mitochondrial Protein | |
Research Use Only | |
MRPS6 | |
Recombinant Protein | |
E. coli | |
PRYELALILK AMQRPETAAT LKRTIEALMD RGAIVRDLEN LGERALPYRI SAHSQQHNRG GYFLVDFYAP TAAVESMVEH LSRDIDVIRG NIVKHPLTQE LKECEGIVPV PLAEKLYSTK KRKK | |
Greater than 80% as determined by SDS-PAGE. |
64968 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |