missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human 28S ribosomal protein S6, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) 28S ribosomal protein S6, mitochondrial, was expressed in the hosts and tags indicated.
245.00€ - 3405.00€
Specifications
Accession Number | NP_115865.1 |
---|---|
Gene ID (Entrez) | 64968 |
Name | 28S ribosomal protein S6, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16042930
|
enQuireBio™
QP6386-EC-100UG |
100 μg |
770.00€
100µg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16062930
|
enQuireBio™
QP6386-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 17-06-2024
Log in to see stock. |
|||||
16052930
|
enQuireBio™
QP6386-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16072930
|
enQuireBio™
QP6386-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16082930
|
enQuireBio™
QP6386-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16092930
|
enQuireBio™
QP6386-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_115865.1 | |
28S ribosomal protein S6, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
64968 | |
Research Use Only | |
MRPS6 | |
Recombinant Protein | |
E. coli | |
PRYELALILK AMQRPETAAT LKRTIEALMD RGAIVRDLEN LGERALPYRI SAHSQQHNRG GYFLVDFYAP TAAVESMVEH LSRDIDVIRG NIVKHPLTQE LKECEGIVPV PLAEKLYSTK KRKK | |
Greater than 80% as determined by SDS-PAGE. |