missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human 14-3-3 eta / YWHAH Protein

A DNA sequence encoding the Homo sapiens (Human) 14-3-3 eta / YWHAH, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP870-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16096011

  • 213.00€ / 10µg

Estimated Shipment: 27-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_003396.1
14-3-3 eta / YWHAH Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
7533
Greater than 90% as determined by SDS-PAGE.
Research Use Only
YWHAH
Recombinant Protein
E. coli
REQLLQRARL AEQAERYDDM ASAMKAVTEL NEPLSNEDRN LLSVAYKNVV GARRSSWRVI SSIEQKTMAD GNEKKLEKVK AYREKIEKEL ETVCNDVLSL LDKFLIKNCN DFQYESKVFY LKMKGDYYRY LAEVASGEKK NSVVEASEAA YKEAFEISKE QMQPTHPIRL GLALNFSVFY YEIQNAPEQA CLLAKQAFDD AIAELDTLNE DSYKDSTLIM QLLRDNLTLW TSDQQDEEAG EGN
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers