missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abcam Recombinant Hepatitis C Virus NS3 Protein, >95% Purity, 100 ug
Description
- Protein Type: Recombinant Hepatitis C Virus NS3 protein (isolate BK)
- Expression System: Escherichia coli
- Amino Acid Range: 1450 to 1643 aa
- Purity: >95%
- Applications: ELISA, Flow Cyt, SDS-PAGE, WB, FuncS
- Formulation: Liquid. pH 7.2 to 7.6. Constituents include 50% glycerol, 48% urea, 0.316% Tris HCl, and 0.078% 2-mercaptoethanol
- Concentration: 1 mg/mL
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 50% Glycerol, 48% urea, 0.316% Tris-HCl, 0.078% 2-mercaptoethanol, pH7.2 to pH7.6 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -20°C. |
| Form | Liquid |
| Label or Dye | Unconjugated |
| Shipping Condition | Blue Ice |
| Target Molecule | Hepatitis C Virus NS3 |
| Formulation | 50% Glycerol, 48% Urea, 0.316% Tris HCl, 0.078% 2-Mercaptoethanol, pH 7.2 to 7.6 |
| Quantity | 100 μg |
| For Use With (Application) | SDS-PAGE, ELISA, Western Blot, Flow Cytometry, FuncS |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?