missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Guinea Pig IFNG / Interferon Gamma Protein

A DNA sequence encoding the Cavia porcellus (Guinea pig) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8811-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16066611

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_001166345.1
IFNG / Interferon Gamma Protein
Research Use Only
IFNG
Recombinant Protein
E. coli
QSRFTNEIRI LKNYFNADNS DVGDNGTLFV GILKNCQEES ERKIFQSQIV SFYFKLFEKH FTDNQTVQNS MNTIKEQIIT KFFKDNSSNK VQAFKNLIQI SVNDEHVQRQ AIIELKKVID DLSPNQRKRR RTQMLFQSRR ASK
Greater than 90% as determined by SDS-PAGE.
100379568
500 μg
Not determined.
Full Length
Guinea Pig
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers