missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Guinea Pig IFNG / Interferon Gamma Protein
A DNA sequence encoding the Cavia porcellus (Guinea pig) IFNG / Interferon Gamma Protein, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_001166345.1 |
---|---|
Gene ID (Entrez) | 100379568 |
Name | IFNG / Interferon Gamma Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16026611
|
enQuireBio™
QP8811-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
16046611
|
enQuireBio™
QP8811-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16036611
|
enQuireBio™
QP8811-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16056611
|
enQuireBio™
QP8811-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16066611
|
enQuireBio™
QP8811-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16076611
|
enQuireBio™
QP8811-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_001166345.1 | |
IFNG / Interferon Gamma Protein | |
Not determined. | |
Full Length | |
Guinea Pig | |
His | |
Tris-based buffer, 50% glycerol |
100379568 | |
Research Use Only | |
IFNG | |
Recombinant Protein | |
E. coli | |
QSRFTNEIRI LKNYFNADNS DVGDNGTLFV GILKNCQEES ERKIFQSQIV SFYFKLFEKH FTDNQTVQNS MNTIKEQIIT KFFKDNSSNK VQAFKNLIQI SVNDEHVQRQ AIIELKKVID DLSPNQRKRR RTQMLFQSRR ASK | |
Greater than 90% as determined by SDS-PAGE. |