missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| Formulation | Lyophilized from a 0.2 μm filtered concentrated solution in PBS, pH 7.4. |
| Molecular Weight (g/mol) | MolecularWeight-theroretical: 33.1 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.) |
| Endotoxin Concentration | Less than 0.1 EU/μg of Flagellin protein FliA(H) as determined by LAL method. |
| Gene Alias | FliA, RNA polymerase sigma factor FliA, Sigma F, Sigma-27, Sigma-28 |
| Gene Symbol | fliA(H) |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.) |
| Purity or Quality Grade | >97 % pure by SDS-PAGE and HPLC |
| Protein | Flagellin protein FliA(H) |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?