missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Feline IL-4 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
Description
The Recombinant Feline IL-4 Protein is derived from E. coli. The Recombinant Feline IL-4 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 12.6 kDa |
| Name | IL-4 |
| Quantity | 25 μg |
| Source | E. coli-derived feline IL-4 protein Gly24-His133 & Asn26-His133 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction