missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr Trans-activator protein BZLF1 Protein

Product Code. p-7153985
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. Quantity unitSize
16007970 10 μg 10µg
16047970 50 μg 50µg
16097960 100 μg 100µg
16027970 200 μg 200µg
16037970 500 μg 500µg
16017970 1 mg 1mg
6 options
This item is not returnable. View return policy

Product Code. 16007970

Brand: enQuireBio™ QP7277ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Trans-activator protein BZLF1, was expressed in the hosts and tags indicated.

Specifications

Accession Number YP_401673.1
Gene ID (Entrez) 3783744
Name Trans-activator protein BZLF1 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol BZLF1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Epstein-Barr
Species E. coli
Protein Tag His-SUMO
Sequence MMDPNSTSED VKFTPDPYQV PFVQAFDQAT RVYQDLGGPS QAPLPCVLWP VLPEPLPQGQ LTAYHVSTAP TGSWFSAPQP APENAYQAYA APQLFPVSDI TQNQQTNQAG GEAPQPGDNS TVQTAAAVVF ACPGANQGQQ LADIGVPQPA PVAAPARRTR KPQQPESLEE CDSELEIKRY KNRVASRKCR AKFKQLLQHY REVAAAKSSE NDRLRLLLKQ MCPSLDVDSI IPRTPDVLHE DLLNF
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.