missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr Apoptosis regulator BHRF1 Protein

Product Code. 16097000
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16097000

Brand: enQuireBio™ QP700510ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Apoptosis regulator BHRF1, was expressed in the hosts and tags indicated.

Specifications

Accession Number YP_401646.1
Gene ID (Entrez) 3783706
Name Apoptosis regulator BHRF1 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol BHRF1
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Epstein-Barr
Species E. coli
Protein Tag His
Sequence MAYSTREILL ALCIRDSRVH GNGTLHPVLE LAARETPLRL SPEDTVVLRY HVLLEEIIER NSETFTETWN RFITHTEHVD LDFNSVFLEI FHRGDPSLGR ALAWMAWCMH ACRTLCCNQS TPYYVVDLSV RGMLEASEGL DG
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.