missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Type-1 fimbrial protein, A chain Protein
A DNA sequence encoding the Escherichia coli (strain K12) Type-1 fimbrial protein, A chain, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7353-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_418734.1 | |
Type-1 fimbrial protein, A chain Protein | |
Research Use Only | |
fimA | |
Recombinant Protein | |
E. coli | |
AATTVNGGTV HFKGEVVNAA CAVDAGSVDQ TVQLGQVRTA SLAQEGATSS AVGFNIQLND CDTNVASKAA VAFLGTAIDA GHTNVLALQS SAAGSATNVG VQILDRTGAA LTLDGATFSS ETTLNNGTNT IPFQARYFAT GAATPGAANA DATFKVQYQ | |
Greater than 80.3% as determined by SDS-PAGE. |
948838 | |
100 μg | |
Not determined. | |
Full Length of Mature Protein | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |