missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Type-1 fimbrial protein, A chain Protein
A DNA sequence encoding the Escherichia coli (strain K12) Type-1 fimbrial protein, A chain, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | NP_418734.1 |
---|---|
Gene ID (Entrez) | 948838 |
Name | Type-1 fimbrial protein, A chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16028390
|
enQuireBio™
QP7353-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16048390
|
enQuireBio™
QP7353-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
16038390
|
enQuireBio™
QP7353-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058390
|
enQuireBio™
QP7353-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16068390
|
enQuireBio™
QP7353-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16078390
|
enQuireBio™
QP7353-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_418734.1 | |
Type-1 fimbrial protein, A chain Protein | |
Not determined. | |
Full Length of Mature Protein | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
948838 | |
Research Use Only | |
fimA | |
Recombinant Protein | |
E. coli | |
AATTVNGGTV HFKGEVVNAA CAVDAGSVDQ TVQLGQVRTA SLAQEGATSS AVGFNIQLND CDTNVASKAA VAFLGTAIDA GHTNVLALQS SAAGSATNVG VQILDRTGAA LTLDGATFSS ETTLNNGTNT IPFQARYFAT GAATPGAANA DATFKVQYQ | |
Greater than 80.3% as determined by SDS-PAGE. |