Learn More
enQuireBio™ Recombinant E. coli tetR Protein
A DNA sequence encoding the Escherichia coli tetR, was expressed in the hosts and tags indicated.
309.00€ - 3405.00€
Specifications
Accession Number | WP_000088605.1 |
---|---|
Gene ID (Entrez) | 31515962 |
Name | tetR Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008770
|
enQuireBio™
QP7416-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||||
16028770
|
enQuireBio™
QP7416-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 11-07-2024
Log in to see stock. |
|||||
16018770
|
enQuireBio™
QP7416-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16038770
|
enQuireBio™
QP7416-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16048770
|
enQuireBio™
QP7416-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16058770
|
enQuireBio™
QP7416-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
WP_000088605.1 | |
tetR Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
31515962 | |
Research Use Only | |
tetR | |
Recombinant Protein | |
E. coli | |
MSRLDKSKVI NSALELLNEV GIEGLTTRKL AQKLGVEQPT LYWHVKNKRA LLDALAIEML DRHHTHFCPL EGESWQDFLR NNAKSFRCAL LSHRDGAKVH LGTRPTEKQY ETLENQLAFL CQQGFSLENA LYALSAVGHF TLGCVLEDQE HQVAKEERET PTTDSMPPLL RQAIELFDHQ GAEPAFLFGL ELIICGLEKQ LKCESGS | |
Greater than 90% as determined by SDS-PAGE. |