missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Oxygen-insensitive NADPH nitroreductase Protein
A DNA sequence encoding the Escherichia coli (strain K12) Oxygen-insensitive NADPH nitroreductase, was expressed in the hosts and tags indicated.
437.00€ - 3405.00€
Specifications
Accession Number | NP_415372.1 |
---|---|
Gene ID (Entrez) | 945483 |
Name | Oxygen-insensitive NADPH nitroreductase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16007350
|
enQuireBio™
QP7087-EC-100UG |
100 μg |
985.00€
100µg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16017350
|
enQuireBio™
QP7087-EC-1MG |
1 mg |
3405.00€
1mg |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||||
16027350
|
enQuireBio™
QP7087-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16037350
|
enQuireBio™
QP7087-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
16047350
|
enQuireBio™
QP7087-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Specifications
NP_415372.1 | |
Oxygen-insensitive NADPH nitroreductase Protein | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
945483 | |
Research Use Only | |
nfsA | |
Recombinant Protein | |
E. coli | |
MTPTIELICG HRSIRHFTDE PISEAQREAI INSARATSSS SFLQCSSIIR ITDKALREEL VTLTGGQKHV AQAAEFWVFC ADFNRHLQIC PDAQLGLAEQ LLLGVVDTAM MAQNALIAAE SLGLGGVYIG GLRNNIEAVT KLLKLPQHVL PLFGLCLGWP ADNPDLKPRL PASILVHENS YQPLDKGALA QYDEQLAEYY LTRGSNNRRD TWSDHIRRTI IKESRPFILD YLHKQGWATR | |
Greater than 90% as determined by SDS-PAGE. |