missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Oxygen-insensitive NADPH nitroreductase Protein

A DNA sequence encoding the Escherichia coli (strain K12) Oxygen-insensitive NADPH nitroreductase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7087-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16047350

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance


Description

Description

Specifications

Specifications

NP_415372.1
Oxygen-insensitive NADPH nitroreductase Protein
Research Use Only
nfsA
Recombinant Protein
E. coli
MTPTIELICG HRSIRHFTDE PISEAQREAI INSARATSSS SFLQCSSIIR ITDKALREEL VTLTGGQKHV AQAAEFWVFC ADFNRHLQIC PDAQLGLAEQ LLLGVVDTAM MAQNALIAAE SLGLGGVYIG GLRNNIEAVT KLLKLPQHVL PLFGLCLGWP ADNPDLKPRL PASILVHENS YQPLDKGALA QYDEQLAEYY LTRGSNNRRD TWSDHIRRTI IKESRPFILD YLHKQGWATR
Greater than 90% as determined by SDS-PAGE.
945483
50 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers