Learn More
enQuireBio™ Recombinant E. coli Murein tetrapeptide carboxypeptidase Protein
Product Code. 16076880
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Product Code. 16076880
Brand: enQuireBio™ QP695610ug
A DNA sequence encoding the Escherichia coli (strain K12) Murein tetrapeptide carboxypeptidase, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_415710.1
Gene ID (Entrez) 945756
Name Murein tetrapeptide carboxypeptidase Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ldcA
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His
Sequence MSLFHLIAPS GYCIKQHAAL RGIQRLTDAG HQVNNVEVIA RRCERFAGTE TERLEDLNSL ARLTTPNTIV LAVRGGYGAS RLLADIDWQA LVARQQHDPL LICGHSDFTA IQCGLLAHGN VITFSGPMLV ANFGADELNA FTEHHFWLAL RNETFTIEWQ GEGPTCRAEG TLWGGNLAML ISLIGTPWMP KIENGILVLE DINEHPFRVE RMLLQLYHAG ILPRQKAIIL GSFSGSTPND YDAGYNLESV YAFLRSRLSI PLITGLDFGH EQRTVTLPLG AHAILNNTRE GTQLTISGHP VLKM
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.