missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Methyl-accepting chemotaxis protein II Protein

Product Code. 16098050
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16098050

Brand: enQuireBio™ QP7296ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) Methyl-accepting chemotaxis protein II, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_416400.1
Gene ID (Entrez) 946399
Name Methyl-accepting chemotaxis protein II Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol tar
Protein Length Cytoplasmic Domain
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His-SUMO
Sequence IRRMLLTPLA KIIAHIREIA GGNLANTLTI DGRSEMGDLA QSVSHMQRSL TDTVTHVREG SDAIYAGTRE IAAGNTDLSS RTEQQASALE ETAASMEQLT ATVKQNADNA RQASQLAQSA SDTAQHGGKV VDGVVKTMHE IADSSKKIAD IISVIDGIAF QTNILALNAA VEAARAGEQG RGFAVVAGEV RNLASRSAQA AKEIKALIED SVSRVDTGSV LVESAGETMN NIVNAVTRVT DIMGEIASAS DEQSRGIDQV ALAVSEMDRV TQQNASLVQE SAAAAAALEE QASRLTQAVS AFRLAASPLT NKPQTPSRPA SEQPPAQPRL RIAEQDPNWE TF
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.