missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli GOR Protein
A cDNA sequence encoding the GOR was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10650-20ug
Additional Details : Weight : 0.01000kg
Specifications
GOR Protein | |
Research Use Only | |
The specific activity is > 52 units/ml. One unit will reduce 1.0 umol of oxidized glutathione per minute at pH 7.5 at 25C. | |
E. coli | |
His | |
The GOR solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.1M NaCl and 1mM DTT. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTKHYDY IAIGGGSGGI ASINRAAMYG QKCALIEAKE LGGTCVNVGC VPKKVMWHAA QIREAIHMYG PDYGFDTTIN KFNWETLIAS RTAYIDRIHT SYENVLGKNN VDVIKGFARF VDAKTLEVNG ETITADHILI ATGGRPSHPD IPGVEYGIDSDGFFALPALP ERVAVVGAGY IAVELAGVIN GLGAKTHLFV RKHAPLRSFD PMISETLVEV MNAEGPQLHT NAIPKAVVKN TDGSLTLELE DGRSETVDCL IWAIGREPAN DNINLEAAGV KTNEKGYIVV DKYQNTNIEG IYAVGDNTGA VELTPVAVAA GRRLSERLFN NKPDEHLDYSNIPTVVFSHP PIGTVGLTEP QAREQYGDDQ VKVYKSSFTA MYTAVTTHRQ PCRMKLVCVG SEEKIVGIHG IGFGMDEMLQ GFAVALKMGA TKKDFDNTVA IHPTAAEEFV TMR | |
Greater than 90.0% as determined by SDS-PAGE. |