missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli GOR Protein
A cDNA sequence encoding the GOR was constructed and used to recombinantly synthesize the protein.
137.00€ - 4090.00€
Specifications
Name | GOR Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The specific activity is > 52 units/ml. One unit will reduce 1.0 umol of oxidized glutathione per minute at pH 7.5 at 25C. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15904528
|
enQuireBio™
QP10650-5UG |
5 μg |
137.00€
5µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15994518
|
enQuireBio™
QP10650-20UG |
20 μg |
212.00€
20µg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
15984518
|
enQuireBio™
QP10650-1MG |
1 mg |
4090.00€
1mg |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||||
Specifications
GOR Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMTKHYDY IAIGGGSGGI ASINRAAMYG QKCALIEAKE LGGTCVNVGC VPKKVMWHAA QIREAIHMYG PDYGFDTTIN KFNWETLIAS RTAYIDRIHT SYENVLGKNN VDVIKGFARF VDAKTLEVNG ETITADHILI ATGGRPSHPD IPGVEYGIDSDGFFALPALP ERVAVVGAGY IAVELAGVIN GLGAKTHLFV RKHAPLRSFD PMISETLVEV MNAEGPQLHT NAIPKAVVKN TDGSLTLELE DGRSETVDCL IWAIGREPAN DNINLEAAGV KTNEKGYIVV DKYQNTNIEG IYAVGDNTGA VELTPVAVAA GRRLSERLFN NKPDEHLDYSNIPTVVFSHP PIGTVGLTEP QAREQYGDDQ VKVYKSSFTA MYTAVTTHRQ PCRMKLVCVG SEEKIVGIHG IGFGMDEMLQ GFAVALKMGA TKKDFDNTVA IHPTAAEEFV TMR | |
Greater than 90.0% as determined by SDS-PAGE. |
Research Use Only | |
The specific activity is > 52 units/ml. One unit will reduce 1.0 umol of oxidized glutathione per minute at pH 7.5 at 25C. | |
E. coli | |
His | |
The GOR solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.1M NaCl and 1mM DTT. |