missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Glutamate-pyruvate aminotransferase AlaC Protein
A DNA sequence encoding the Escherichia coli (strain K12) Glutamate-pyruvate aminotransferase AlaC, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6950-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416880.1 | |
Glutamate-pyruvate aminotransferase AlaC Protein | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
946850 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
alaC | |
Recombinant Protein | |
E. coli | |
MADTRPERRF TRIDRLPPYV FNITAELKMA ARRRGEDIID FSMGNPDGAT PPHIVEKLCT VAQRPDTHGY STSRGIPRLR RAISRWYQDR YDVEIDPESE AIVTIGSKEG LAHLMLATLD HGDTVLVPNP SYPIHIYGAV IAGAQVRSVP LVEGVDFFNE LERAIRESYP KPKMMILGFP SNPTAQCVEL EFFEKVVALA KRYDVLVVHD LAYADIVYDG WKAPSIMQVP GARDVAVEFF TLSKSYNMAG WRIGFMVGNK TLVSALARIK SYHDYGTFTP LQVAAIAALE GDQQCVRDIA EQYKRRRDVL VKGLHEAGWM VEMPKASMYV WAKIPEPYAA MGSLEFAKKL LNEAKVCVSP GIGFGDYGDT HVRFALIENR DRIRQAIRGI KAMFRADGLL PASSKHIHEN AE |